11-Aminoundecanoic acid, Sigma-Aldrich®
Catalog # 8.01228.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:2 g/l (20 °C)
- Melting Point:185 °C
- Molar Mass:201.3 g/mol
- Bulk Density:500 kg/m3
Specification Test Results
About this item
:
:201.31 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:187…190 °C
:M-Clarity™ quality level = MQ200
EINECS:219-417-6
CAS Number:2432-99-7
Formula:C₁₁H₂₃NO₂
MDL Number:MFCD00008150