
1-Ethyl-3-methylimidazolium chloride, Sigma-Aldrich®
Catalog # 4.90054.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Melting Point:77 - 79 °C
- Molar Mass:146.62 g/mol
Specification Test Results
About this item
:
:146.62 g/mol
Melting Pt:∼80 °C
Density:∼1,1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:186 °C
:GHS07
CAS Number:65039-09-0
Formula:C₆H₁₁ClN₂
MDL Number:MFCD00074843