Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
6-Propyl-2-thiouracil, Supelco®
Catalog # 46698-250MG
Supplier:  Merck
CAS Number:  
undefined
Irritant
6-Propyl-2-thiouracil, Supelco®
Catalog # 46698-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :170.24 g/mol
Merck Index:12,08054
Boiling Pt:448 °C (1013 hPa)
Melting Pt:215…218 °C
 :M-Clarity™ quality level = MQ200
EINECS:200-103-2
 :GHS07
CAS Number:51-52-5
Formula:C₇H₁₀N₂OS
MDL Number:MFCD00006041