Order Entry
Northern Ireland
ContactUsLinkComponent
 
Nitric acid 70% ≥70%, CMOS for microelectronic, J.T.Baker®
Catalog # 9606-03
Supplier:  Avantor
CAS Number:  
undefined
Nitric acid 70% ≥70%, CMOS for microelectronic, J.T.Baker®
Catalog # 9606-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, PVC-coated
  • Pk:
    7 lb
  • Seq:
    7068
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (HNO₃}:
    69.0 - 70.0 %
  • Appearance Passes Test:
    Passes Test
  • Color (APHA) :
    ≤10
  • Residue after Ignition :
    ≤2 ppm
  • Chloride (Cl) :
    ≤0.08 ppm
  • Phosphate (PO₄) :
    ≤0.1 ppm
  • Sulfate (SO₄) :
    ≤0.2 ppm
  • Trace Impurities - Aluminum (Al) :
    ≤40.0 ppb
  • Arsenic and Antimony (as As) :
    ≤5 ppb
  • Trace Impurities - Barium (Ba) :
    ≤10.0 ppb
  • Trace Impurities - Beryllium (Be) :
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi) :
    ≤20.0 ppb
  • Trace Impurities - Boron (B) :
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd) :
    ≤50.0 ppb
  • Trace Impurities - Calcium (Ca) :
    ≤50.0 ppb
  • Trace Impurities - Chromium (Cr) :
    ≤30.0 ppb
  • Trace Impurities - Cobalt (Co) :
    ≤10.0 ppb
  • Trace Impurities - Copper (Cu) :
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga) :
    ≤10.0 ppb
  • Trace Impurities - Germanium (Ge) :
    ≤20.0 ppb
  • Trace Impurities - Gold (Au) :
    ≤20.0 ppb
  • Heavy Metals (as Pb) :
    ≤100 ppb
  • Trace Impurities - Iron (Fe) :
    ≤40.0 ppb
  • Trace Impurities - Lead (Pb) :
    ≤20.0 ppb
  • Trace Impurities - Lithium (Li) :
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg) :
    ≤20.0 ppb
  • Trace Impurities - Manganese (Mn) :
    ≤10.0 ppb
  • Trace Impurities - Potassium (K) :
    ≤50.0 ppb
  • Trace Impurities - Silicon (Si) :
    ≤50.0 ppb
  • Trace Impurities - Silver (Ag) :
    ≤20.0 ppb
  • Trace Impurities - Sodium (Na) :
    ≤150.0 ppb
  • Trace Impurities - Strontium (Sr) :
    ≤30.0 ppb
  • Trace Impurities - Tantalum (Ta) :
    ≤10.0 ppb
  • Trace Impurities - Thallium (Tl) :
    ≤10.0 ppb
  • Trace Impurities - Tin (Sn) :
    ≤20.0 ppb
  • Trace Impurities - Titanium (Ti) :
    ≤10.0 ppb
  • Trace Impurities - Vanadium (V) :
    ≤10.0 ppb
  • Trace Impurities - Zinc (Zn) :
    ≤20.0 ppb
  • Trace Impurities - Zirconium (Zr) :
    ≤10.0 ppb
  • Particle Count - 0.5 µm and greater :
    ≤60 par/ml
  • Particle Count - 1.0 µm and greater :
    ≤10 par/ml

Specification Test Results

About this item

Synonyms: Nitric acid solution
 :
 :63.01 g/mol
Melting Pt:-42
EINECS:231-714-2
UN:2031
 :GHS03
 :GHS05
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Boiling Pt:87
Density:1.51
ADR:8 I
Merck Index:13,06610