

3-Chloro-4-fluoro-5-nitrobenzotrifluoride ≥98%
Catalog # L19322.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:101646-02-0
:
Formula:C₇H₂ClF₄NO₂
MDL Number:MFCD00674108
:243.54 g/mol
Boiling Pt:200 °C
Density:1,61
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
:GHS07