
1-Butyl-3-methylimidazoliumtetrafluoroborate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:174501-65-6
:
Formula:C₈H₁₅BF₄N₂
MDL Number:MFCD03095449
:226.02 g/mol
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:–71 °C
Density:1,210
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together