5-Amino-3,4-dimethylisoxazole 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19947-75-2
:
Formula:C₅H₈N₂O
MDL Number:MFCD00046081
:112.13 g/mol
Melting Pt:118…120 °C
Storage Temperature:Ambient
EINECS:243-437-4
Frequently Bought Together