
3-(4-Fluorophenyl)propionic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:459-31-4
:
Formula:C₉H₉FO₂
MDL Number:MFCD00060327
:168.17 g/mol
Melting Pt:88…92 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together