2-(4-Aminophenyl)ethanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-10-9
:
Formula:C₈H₁₁NO
MDL Number:MFCD00007922
:137.18 g/mol
Boiling Pt:235…255 °C (1013 hPa)
Melting Pt:108…110 °C
Storage Temperature:Ambient
EINECS:203-174-8
Related Information
Frequently Bought Together