5-(Trifluoromethyl)-o-anisidine ≥97%
Catalog # L08021.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:349-65-5
:
Formula:C₈H₈F₃NO
MDL Number:MFCD00042486
:191.15 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:59 °C
UN:2811
ADR:6.1,III