Order Entry
Northern Ireland
ContactUsLinkComponent
1,1-Dimethylurea ≥98%
1,1-Dimethylurea ≥98%
Catalog # L05065.09
CAS Number:  
1,1-Dimethylurea ≥98%
Catalog # L05065.09
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :88.11 g/mol
Melting Pt:178…183 °C
Density:1,255
EINECS:209-957-0
 :GHS07
Storage Temperature:Ambient
CAS Number:598-94-7
Formula:C₃H₈N₂O
MDL Number:MFCD00007959