

2,3-Dimethyl-1,3-butadiene ≥98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:513-81-5
:
Formula:C₆H₁₀
MDL Number:MFCD00008595
:82.15 g/mol
Merck Index:14,3238
Boiling Pt:68…69 °C
Melting Pt:–76 °C
Density:0,726
Flash Pt:–1 °C (30 °F)
UN:3295
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 ppm
EINECS:208-172-0
:GHS02
:GHS08
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together