di-Sodium β-glycerophosphate pentahydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13408-09-8
:819-83-0
Formula:C₃H₇Na₂O₆P·5H₂O
MDL Number:MFCD00002135
:306.11 g/mol
Merck Index:14,8622
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:212-464-3
Frequently Bought Together