Allylthiourea ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-57-9
:
Formula:H₂CCHCH₂NHC(S)NH₂
MDL Number:MFCD00004940
:116.19 g/mol
Merck Index:14,9362
Boiling Pt:191 °C (1013 hPa)
Melting Pt:70…78 °C
Density:1,220
Storage Temperature:Ambient
EINECS:203-683-5
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together