

4-Fluoro-2-(trifluoromethyl)phenol 95%
Catalog # H31716.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:130047-19-7
:
Formula:C₇H₄F₄O
:180.1 g/mol
Melting Pt:50…54 °C
UN:3261
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00061290