
1-Ethyl-3-methylimidazolium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65039-09-0
:
Formula:C₆H₁₁ClN₂
MDL Number:MFCD00074843
:146.62 g/mol
Melting Pt:77…80 °C
Density:∼1,1 g/cm³ (20 °C)
Flash Pt:186 °C (367 °F)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together