
1-Ethyl-3-methylimidazolium ethylsulphate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:342573-75-5
:
Formula:C₈H₁₆N₂O₄S
MDL Number:MFCD06798189
:236.29 g/mol
Melting Pt:< –30 °C
Density:1,24
Flash Pt:162 °C (324 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together