

(1-Butyl)triphenylphosphonium chloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13371-17-0
:
Formula:H₃C(CH₂)₃P(C₆H₅)₃Cl
MDL Number:MFCD00040546
:354.86 g/mol
Melting Pt:225…228 °C
Flash Pt:>270 °C
Storage Temperature:Ambient
EINECS:236-443-3
UN:3464
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together