2-Bromo-6-(trifluoromethyl)pyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:189278-27-1
:
Formula:C₆H₃BrF₃N
MDL Number:MFCD00153087
:226 g/mol
Boiling Pt:78…79 °C (14 mmHg)
Melting Pt:48…51 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together