

(Benzyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1449-46-3
:
Formula:C₂₅H₂₂BrP
MDL Number:MFCD00031707
Melting Pt:293…298 °C
Storage Temperature:Ambient
EINECS:215-908-4
UN:3464
ADR:6.1,II
:433.33 g/mol
:GHS06
:GHS07
Frequently Bought Together