
N-Isopropylbenzylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102-97-6
:
Formula:C₆H₅CH₂N(CH₃)(CH₂CH₃)
MDL Number:MFCD00008863
:149.24 g/mol
Boiling Pt:199…201 °C
Melting Pt:∼ 0 °C
Density:0,910
Flash Pt:87 °C (190 °F)
Storage Temperature:Ambient
EINECS:203-067-6
UN:2735
ADR:8,III
:GHS07
Frequently Bought Together