

4-(Trifluoromethyl)pyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3796-24-5
:
Formula:C₆H₄F₃N
MDL Number:MFCD00153289
Boiling Pt:107…110 °C
Density:1,270
Flash Pt:20 °C (68 °F)
ADR:3,II
:147.1 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together