

2-(Trimethylsilyl)ethanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2916-68-9
:
Formula:HOH₂CCH₂Si(CH₃)₃
MDL Number:MFCD00002825
:118.25 g/mol
Boiling Pt:71…72 °C (10 mmHg)
Melting Pt:0 °C
Density:0,825
Flash Pt:50 °C (122 °F)
Storage Temperature:Ambient
EINECS:220-844-5
UN:1987
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together