Order Entry
Northern Ireland
ContactUsLinkComponent
(Trifluoromethyl)trimethylsilane ≥98%
Flammable
Irritant
(Trifluoromethyl)trimethylsilane ≥98%
Catalog # B20347.06
CAS Number:  
Flammable
Irritant
(Trifluoromethyl)trimethylsilane ≥98%
Catalog # B20347.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :142.2 g/mol
Boiling Pt:54…55 °C
Density:0,962
Flash Pt:–10 °C (14 °F)
ADR:3,II
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:81290-20-2
Formula:C₄H₉F₃Si
MDL Number:MFCD00145454