Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

2',4'-Dihydroxy-3'-methylacetophenone ≥98%

2',4'-Dihydroxy-3'-methylacetophenone ≥98%
Certificates
2',4'-Dihydroxy-3'-methylacetophenone ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 10139-84-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 152-156° MDL No.: MFCD00010817

Synonyms:  1-(2,4-Dihydroxy-3-methylphenyl)ethanone, 1-(2,4-Dihydroxy-3-methylphenyl)ethan-1-one
  • CAS Number:10139-84-1
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00010817
  • Molecular Weight:166.18 g/mol
  • Melting Pt:152…156 °C
  • Storage Temperature:Ambient
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £59.60
    £59.60
    1 * 5 g
       £59.60

    Specifications:
    More
    Specifications
    Cat. No.
    B20272.06
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £213.00
    £213.00
    1 * 25 g
       £213.00

    Specifications:
    More
    Specifications
    Cat. No.
    B20272.14