Undec-10-enoic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-38-9
:
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00004442
:184.28 g/mol
Merck Index:14,9848
Boiling Pt:126…128 °C (1 mmHg)
Melting Pt:22…25 °C
Density:0,910
Flash Pt:160 °C (320 °F)
EINECS:203-965-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together