Methyl crotonate all trans 96% (by GC), liquid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:623-43-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00009287
:100.12 g/mol
Merck Index:14,2597
Boiling Pt:118…120 °C
Density:0,944
Flash Pt:4 °C (40 °F)
Storage Temperature:Ambient
EINECS:210-793-7
UN:3272
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together