
4-Methoxybenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-68-0
:
Formula:C₇H₇ClO₃S
MDL Number:MFCD00007446
Boiling Pt:119…122 °C (0,4 mmHg)
Melting Pt:37…42 °C
Flash Pt:>110 °C (230 °F)
EINECS:202-692-1
UN:3261
ADR:8,II
:206.65 g/mol
:GHS05
Related Information
Frequently Bought Together