
Tris(4-chlorophenyl)phosphine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1159-54-2
:
Formula:C₁₈H₁₂Cl₃P
MDL Number:MFCD00013639
:365.63 g/mol
Boiling Pt:431 °C (1013 hPa)
Melting Pt:102…103 °C
Density:1,449 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-596-7
:GHS07
Related Information
Frequently Bought Together