

Titanium(IV) butanolate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5593-70-4
:
Formula:C₁₆H₃₆O₄Ti
MDL Number:MFCD00009433
:340.34 g/mol
Boiling Pt:310…314 °C
Melting Pt:–55 °C
Density:0,998
Flash Pt:41 °C (170 °F)
Storage Temperature:Ambient
EINECS:227-006-8
UN:2924
ADR:3,III
:GHS05
:GHS02
Related Information
Certificates
Search by Lot NumberThermo Scientific Organic Photovoltaics Thermo Scientific Organometallic
Product Literature
Frequently Bought Together