

1-Butyl-3-methylimidazolium chloride 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79917-90-1
:
Formula:C₈H₁₅ClN₂
MDL Number:MFCD03095425
:174.67 g/mol
Boiling Pt:? 200 °C (1013 hPa)
Melting Pt:73 °C
Density:~ 1 g/cm³ (20 °C)
Flash Pt:≥ 110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:Cream to yellow to orange Powder or crystals or chunks or fused solid
- Infrared spectrum:Conforms
- Water:≤1.0 % (K.F.)
- HPLC:≥98.0 %
Specification Test Results
Frequently Bought Together