

4-Chloro-o-phenylenediamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-83-0
:
Formula:C₆H₇ClN₂
MDL Number:MFCD00011691
:142.59 g/mol
Boiling Pt:280 °C (1013 hPa)
Melting Pt:70…75 °C
EINECS:202-456-8
:GHS07
:GHS08
Specification Test Results
- Appearance:Grey-brown to dark brown Powder or granules and/or chunks
- Infrared spectrum:Conforms
- Melting point:70 to 75 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together