
Sodium nitroprusside dihydrate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13755-38-9
:
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192
Merck Index:13,08722
Density:1,72 g/cm³
Storage Temperature:Ambient
EINECS:238-373-9
UN:1588
ADR:6.1,III
:297.95 g/mol
:GHS06
Specification Test Results
- Appearance:Red to dark red-brown Powder or crystals and/or chunks
- Titration Argentometric:≥99.0 %
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤100 ppm
- Hexacyanoferrate III:≤0.01 %
- Hexacyanoferrate II:≤0.02 %
Specification Test Results
Frequently Bought Together