
2-(4-Methylthiazol-5-yl)ethanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:137-00-8
:
Formula:C₆H₉NOS
MDL Number:MFCD00005339
:143.21 g/mol
Merck Index:12,06208
Boiling Pt:135 °C (7 mmHg)
Density:1,19 g/cm³
Flash Pt:>110 °C
EINECS:205-272-6
:GHS07
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5480 to 1.5510 (20°C, 589 nm)
- Water:≤1 %
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together