Order Entry
Northern Ireland
ContactUsLinkComponent
Undec-10-enoyl chloride 98%
Undec-10-enoyl chloride 98%
Certificates
undefined
Undec-10-enoyl chloride 98%

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4532 to 1.4552 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together