ContactUsLinkComponent
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
Certificates
undefined
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate 98%
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Complexometric:
    97.5 to 102.5 %
  • Loss on drying:
    4.0 to 6.9 % (1g,150°C,6h)
  • Sulfated ash:
    ≤0.3 %