Order Entry
Northern Ireland
ContactUsLinkComponent
Propiolic acid 98%
Flammable
Corrosive
Toxic
Propiolic acid 98%
Certificates
undefined
Flammable
Corrosive
Toxic
Propiolic acid 98%

Specification Test Results

  • Appearance:
    Clear colorless to brown Liquid or solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1,4305 to 1,4345 (20 °C, 589 nm)
  • Titration with NaOH:
    97,5 to 102,5 %

Specification Test Results

Related Information

Frequently Bought Together