Order Entry
Northern Ireland
ContactUsLinkComponent
N-Phenyl-o-phenylenediamine 97%
Irritant
Health
N-Phenyl-o-phenylenediamine 97%
Catalog # ACRO130770500
CAS Number:  
undefined
Irritant
Health
N-Phenyl-o-phenylenediamine 97%
Catalog # ACRO130770500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Appearance:
    Pink to grey-brown to violet Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    77 to 82 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :184.24 g/mol
Boiling Pt:106…108 °C (0,4 mmHg)
Melting Pt:77…82 °C
Density:1,234 g/cm³ (20 °C)
EINECS:208-606-9
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:534-85-0
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00007685