1-Methyl-2-thiourea 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:598-52-7
:
Formula:C₂H₆N₂S
MDL Number:MFCD00004938
Melting Pt:118…123 °C
EINECS:209-936-6
UN:2811
ADR:6.1,II
:90.15 g/mol
:GHS06
:GHS08
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:118 to 123 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together