Indazole 96%
:271-44-3
:
:C₇H₆N₂
:MFCD00005691
:118.14 g/mol
:13,04961
:270 °C (743 mmHg)
:146…150 °C
:1,32 g/cm³ (20 °C)
:Ambient
:205-978-4
- Appearance:White to beige Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:146 to 150 °C
- Titration with HClO4:≥95 %
Frequently Bought Together