11-Aminoundecanoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2432-99-7
:
Formula:C₁₁H₂₃NO₂
MDL Number:MFCD00008150
:201.31 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:187…190 °C
EINECS:219-417-6
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:187 to 190 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together