Order Entry
Northern Ireland
ContactUsLinkComponent
 
L(-)-Proline
Catalog # MOLE90004692
Supplier:  Molekula
CAS Number:  
L(-)-Proline
Catalog # MOLE90004692
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :115.13 g/mol
Merck Index:13,07871
Boiling Pt:245 °C (1013 hPa)
Melting Pt:220…222 °C
Density:1,35 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:205-702-2
CAS Number:147-85-3
Formula:C₅H₉NO₂
MDL Number:MFCD00064318