ContactUsLinkComponent
Amplite® Red Peroxidase Substrate
Amplite® Red Peroxidase Substrate
Catalog # AATB11011
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Red Peroxidase Substrate
Catalog # AATB11011
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    1.000 Assays
  • Description:
    Amplite™ Red
Specifications
About this item
Amplite® Red is a sensitive fluorogenic peroxidase substrate that generates a highly red fluorescent product that has maximum absorption of 571 nm and maximum emission of 585 nm.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.