ContactUsLinkComponent
Amplite® Ethanol Quantitation Kit
Amplite® Ethanol Quantitation Kit
Catalog # AATB40001
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Ethanol Quantitation Kit
Catalog # AATB40001
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ ethanol quantitation kit
Specifications
About this item
The ability to rapidly perform quantitative measurements of ethanol is highly desirable in life science research, clinical evaluations, and food and drug industries.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.