Iron ≥99.5%, spherical powder APS 6-10 µm,, reduced
10214.95
:
- Pack type:Glass bottle for solids
- Pk:2
- Seq:0002
- Environmentally Preferable:
:
:55.85 g/mol
:1535
:231-096-4
:3089
:GHS02
:GHS07
:Ambient
:7439-89-6
:Fe
:MFCD00010999
:3000
:7.86 g/cm³ (20 °C)
:4.1 II
:13,05109