Order Entry
Northern Ireland
ContactUsLinkComponent
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates
undefined
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test
  • Formaldehyde:
    Max. 0.1 %
  • Methanol:
    Max. 0.1 %
  • Water:
    Max. 0.05 %

Specification Test Results

Frequently Bought Together