Order Entry
Northern Ireland
ContactUsLinkComponent
Fluorescein disodium salt ≥95%
Fluorescein disodium salt ≥95%
Catalog # AATB2
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Fluorescein disodium salt ≥95%
Catalog # AATB2
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate
 :
 :376.28 g/mol
 :GHS07
CAS Number:518-47-8
Formula:C₂₀H₁₀Na₂O₅
MDL Number:MFCD00167039
ADR:5 II
Merck Index:13,04185