Order Entry
Northern Ireland
ContactUsLinkComponent
SPINeasy® DNA Kit for Blood, MP Biomedicals
SPINeasy® DNA Kit for Blood, MP Biomedicals
Catalog # 116552050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® DNA Kit for Blood, MP Biomedicals
Catalog # 116552050.
Supplier:  MP Biomedicals
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Saliva
    Blood
    Cell culture
    Plasma
    Serum
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA kit for blood, 50 tests
  • Storage temperature:
    Store at room temperature (15...25 ℃) in a dry environment.

Specifications

About this item

SPINeasy DNA kit for blood is a high-performance genomic DNA (gDNA) extraction kit based on silica-membrane spin-column technology. This kit enables quick isolation of gDNA from whole blood preserved in different anticoagulants (EDTA, Heparin and sodium citrate), typically in less than 30 minutes.

  • Rapid and efficient DNA isolation in less than 30 minutes
  • Fresh, frozen, or anticoagulated blood
  • Simple and effective lysis protocol using Lysis buffer BL and Proteinase K to omit the need of any mechanical lysis
  • No organic extraction
  • High purity DNA ready to use for downstream application

It also allows easy gDNA isolation from other samples, including plasma, serum, saliva, and cell culture medium. The use of our specially formulated Lysis buffer BL omits the need of homogenization and enables highly efficient lysis of various samples. Provided in the kit, Column S and kit buffers are designed to deliver gDNA extracts of high yield and purity, compatible with downstream applications such as PCR amplification.

Key applications are Genomics; Molecular biology; PCR, qPCR, Microbiome analysis, NGS; shotgun sequencing.

Sample colume 200 μl of Human blood, serum, plasma, saliva, or cell culture media.

Delivery: Kit includes Lysis buffer BL, Wash buffer BW1, Wash buffer BW2, Elution buffer EB, Column S, 2,0 ml collection tubes, 1,5 ml microcentrifuge tubes, Proteinase K, Proteinase K buffer.