Order Entry
Northern Ireland
ContactUsLinkComponent
SPINeasy® DNA Kit for Saliva, MP Biomedicals
SPINeasy® DNA Kit for Saliva, MP Biomedicals
Catalog # 116551050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® DNA Kit for Saliva, MP Biomedicals
Catalog # 116551050.
Supplier:  MP Biomedicals
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Saliva
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA kit for saliva, 50 tests
  • Storage temperature:
    Store at room temperature (15...25 ℃) in a dry environment.

Specifications

About this item

The SPINeasy DNA kit for saliva from MP Biomedicals makes saliva sampling easier by using our specially formulated Saliva Preservation Solution (SPS) to preserve the sample at room temperature without compromising the quality.

  • Specialized saliva preservative solution to store saliva samples at room temperature for a prolonged period
  • Simple and effective extraction protocol with high DNA yield and purity
  • Isolate gDNA from 25 to 500 µl of saliva with simple procedures
  • DNA ready to use for downstream application

The use of saliva as a source of DNA over blood samples has become an attractive approach for various applications ranging from genetic studies to pathogen detection. Unlike blood sampling, saliva collection is an easy, painless, and non-invasive method which do not require trained personnel for the collection process. In addition, saliva does not coagulate which makes it easier for handling than blood. However, storing saliva samples can be problematic.

SPINeasy DNA kit for saliva allows DNA from fresh, frozen, or SPS-preserved saliva to be extracted with a quick and easy protocol, using silica spin-column technology. Purified DNA is recovered in high yield and purity, suitable for various downstream molecular applications.

Delivery: Kit includes Saliva Preservative Solution (SPS), Lysing Matrix Y, Lysis Buffer SL, Inhibitor Remover SL, Binding Buffer SL, Wash Buffer SL1, Wash Buffer SL2, Elution Buffer SL, Column S, 2,0 ml Collection Tubes, 2 ml microcentrifuge tubes, 1,5 ml microcentrifuge tubes.