Order Entry
Northern Ireland
ContactUsLinkComponent
SPINeasy® DNA Extraction Kit for Plant, MP Biomedicals
SPINeasy® DNA Extraction Kit for Plant, MP Biomedicals
Catalog # 116535050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® DNA Extraction Kit for Plant, MP Biomedicals
Catalog # 116535050.
Supplier:  MP Biomedicals
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Extraction for direct PCR
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA extraction kit for plant, 50 tests
  • Storage temperature:
    Store at room temperature (15...30 °C), in a dry environment.

Specifications

About this item

SPINeasy DNA Kit for plant is a silica-membrane spin-column kit that enables quick, convenient and cost-effective isolation of genomic DNA (gDNA) from various plant tissues, such as leaf, stem and seed.

High quality gDNA can be extracted from polysaccharide and polyphenolic-rich plants. The use of our specially formulated Lysis Buffer PD and Lysing Matrix A in combination with FastPrep® Instruments from MP Biomedicals enables highly efficient lysis of various types of samples within seconds. For DNA purification, Column PD and kit buffers are designed to deliver gDNA extracts of high yield and purity which are compatible with downstream molecular biology applications such as PCR, restriction digestion and sequencing.

Key applications are plant genomics, molecular biology, PCR, NGS.

Certifications: Hazard Class 8
Delivery: Provided with 50×2 ml Lysing Matrix A, 50 ml Lysis Buffer PD, 12 ml PPS Solution, 15 ml Binding Buffer PD, 2×12 ml Wash Buffer, 10 ml Elution Buffer PD, 0,8 ml RNase A, 50×Column PD with collection tubes.